4-(N,N-Dimethylamino)phenylboronic acid hydrochloride 98%

Supplier: Ambeed
Warning

A122310-1G A122310-5G A122310-10G A122310-25G
76995-908EA 17 USD
76995-908 77549-436 77670-734 77880-989
4-(N,N-Dimethylamino)phenylboronic acid hydrochloride 98%
4-(N,N-Dimethylamino)phenylboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃BClNO₂
MDL Number: MFCD04112547
CAS Number: 1150114-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR