1-Boc-2-formylpyrrolidine 97%

Supplier: Ambeed
Warning

A122299-1G A122299-5G A122299-10G
77026-490EA 9 USD
77026-490 77566-304 77666-754
1-Boc-2-formylpyrrolidine 97%
1-Boc-2-formylpyrrolidine

Precaución: For research use only.

Formula: C₁₀H₁₇NO₃
MDL Number: MFCD03790896
CAS Number: 117625-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR