2-Acetoxy-2',4'-difluoroacetophenone 97%
Supplier: Ambeed
|
Warning
|
A122256-1G
A122256-250MG
77026-480EA
11
USD
77026-480
77026-482
2-Acetoxy-2',4'-difluoroacetophenone 97%
2-Acetoxy-2',4'-difluoroacetophenone
Precaución: for Research Use Only
Formula:
C₁₀H₈F₂O₃ |
MDL Number:
MFCD09743655 CAS Number: 122263-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...