4-Amino-3-bromo-2-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-methoxypyridin-4-amine, 3-Bromo-2-methoxy-4-pyridinamine

A122251-250MG A122251-5G A122251-1G
76995-904EA 24.41 USD
76995-904 76995-906 77592-384
4-Amino-3-bromo-2-methoxypyridine 98%
4-Amino-3-bromo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MW: 203.0366 g/mol
MDL Number: MFCD11656225
CAS Number: 215364-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR