6-Chloroquinazoline-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A122130-100MG A122130-1G
77026-470EA 6 USD
77026-470 77026-472
6-Chloroquinazoline-2,4(1H,3H)-dione 95%
6-Chloroquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD00030678
CAS Number: 1640-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR