2-Allyl-4,5-dimethoxyphenol 95%

Supplier: Ambeed
Warning

A121984-1G A121984-250MG
76983-316EA 448.77 USD
76983-316 76983-318
2-Allyl-4,5-dimethoxyphenol 95%
2-Allyl-4,5-dimethoxyphenol

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD18975114
CAS Number: 59893-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR