6,6'-Dicyano-2,2'-bipyridyl 97%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Bipyridine-6,6'-dicarbonitrile, [2,2'-Bipyridine]-6,6'-dicarbonitrile

A121980-1G A121980-250MG A121980-5G
76983-310EA 140.04 USD
76983-310 76983-312 76983-314
6,6'-Dicyano-2,2'-bipyridyl 97%
6,6'-Dicyano-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₂H₆N₄
MW: 206.21 g/mol
MDL Number: MFCD00797251
CAS Number: 4411-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR