3-Chloro-4-fluorocinnamic acid 97%

Supplier: Ambeed
Warning

A121960-1G A121960-5G
77026-464EA 18 USD
77026-464 77026-466
3-Chloro-4-fluorocinnamic acid 97%
3-Chloro-4-fluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆ClFO₂
MDL Number: MFCD01631393
CAS Number: 155814-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR