8-Aminoquinoline-7-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 8-Aminoquinoline-7-carbaldehyde

A121925-100MG A121925-1G A121925-250MG
77026-448EA 78.51 USD
77026-448 77026-450 77026-452
8-Aminoquinoline-7-carboxaldehyde 95%
8-Aminoquinoline-7-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.183 g/mol
MDL Number: MFCD11110554
CAS Number: 158753-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR