Benzyl (1,3-dihydroxypropan-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A121899-10G A121899-1G A121899-250MG A121899-25G A121899-5G
76995-868EA 88 USD
76995-868 76995-870 76995-872 76995-874 76995-876
Benzyl (1,3-dihydroxypropan-2-yl)carbamate 97%
Benzyl (1,3-dihydroxypropan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₄
MW: 225.24 g/mol
MDL Number: MFCD11656821
CAS Number: 71811-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR