2-Amino-3-bromo-6-chloropyrazine 98%

Supplier: Ambeed
Warning

A121865-100G A121865-100MG A121865-10G A121865-1G A121865-250MG A121865-25G A121865-5G
76995-854EA 482 USD
76995-854 76995-856 76995-858 76995-860 76995-862 76995-864 76995-866
2-Amino-3-bromo-6-chloropyrazine 98%
2-Amino-3-bromo-6-chloropyrazine

Precaución: for Research Use Only

Formula: C₄H₃BrClN₃
MDL Number: MFCD11044158
CAS Number: 212779-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR