1-Cyclopropyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

A121856-1G A121856-250MG
76983-302EA 14.86 USD
76983-302 76983-304
1-Cyclopropyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylic acid 95%
1-Cyclopropyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₈F₃NO₃
MW: 283.203 g/mol
MDL Number: MFCD01173812
CAS Number: 94695-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR