5-tert-Butyl-[1,2,4]oxadiazole-3-carboxylic acid ethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-(tert-butyl)-1,2,4-oxadiazole-3-carboxylate

A121819-100MG A121819-250MG A121819-5G A121819-1G
77026-442EA 48.81 USD
77026-442 77026-444 77026-446 77666-748
5-tert-Butyl-[1,2,4]oxadiazole-3-carboxylic acid ethyl ester 97%
5-tert-Butyl-[1,2,4]oxadiazole-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₃
MW: 198.219 g/mol
MDL Number: MFCD11975623
CAS Number: 158154-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR