5-Bromo-2-ethylbenzoxazole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-ethylbenzo[d]oxazole

A121622-1G A121622-250MG
76995-838EA 45.62 USD
76995-838 76995-840
5-Bromo-2-ethylbenzoxazole 98%
5-Bromo-2-ethylbenzoxazole

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.0700 g/mol
MDL Number: MFCD09055180
CAS Number: 938458-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR