2-(3-Oxocyclopentyl)isoindoline-1,3-dione 95%
Supplier: Ambeed
|
Warning
|
A121585-10G
A121585-1G
A121585-250MG
A121585-5G
76983-282EA
437.1
USD
76983-282
76983-284
76983-286
76983-288
2-(3-Oxocyclopentyl)isoindoline-1,3-dione 95%
2-(3-Oxocyclopentyl)isoindoline-1,3-dione
Precaución: for Research Use Only
Formula:
C₁₃H₁₁NO₃ MW: 229.23 g/mol |
MDL Number:
MFCD18910252 CAS Number: 1029691-06-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...