Methyl 3,6-dichloropyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

A121531-100MG A121531-1G A121531-250MG
76983-276EA 7 USD
76983-276 76983-278 76983-280
Methyl 3,6-dichloropyrazine-2-carboxylate 97%
Methyl 3,6-dichloropyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
MDL Number: MFCD10697748
CAS Number: 356783-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR