4,5,6-Trichloropyrimidin-2-amine 98%

Supplier: Ambeed
Warning

A121493-100MG A121493-250MG
77026-426EA 38.2 USD
77026-426 77026-428
4,5,6-Trichloropyrimidin-2-amine 98%
4,5,6-Trichloropyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₄H₂Cl₃N₃
MW: 198.44 g/mol
MDL Number: MFCD12405318
CAS Number: 51501-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR