Triacetyl resveratrol 98%

Supplier: Ambeed
Danger

Synonyms: (E)-5-(4-Acetoxystyryl)-1,3-phenylene diacetate, 3,5,4′-Tri-O-acetylresveratrol, Acetyl-trans-resveratrol, 5-[(1E)-2-[4-(acetyloxy)phenyl]ethenyl]-1,3-Benzenediol-1,3-diacetate

A121434-1G A121434-50MG A121434-10G A121434-25G A121434-5G
77026-422EA 6.37 USD
77026-422 77026-424 77666-740 77666-742 77666-744
Triacetyl resveratrol 98%
Triacetyl resveratrol

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₆
MW: 354.35 g/mol
MDL Number: MFCD01546481
CAS Number: 42206-94-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR