2,5-Dichlorothiazole-4-carbonitrile 98%

Supplier: Ambeed
Warning

A121288-1G A121288-250MG
76983-272EA 484.84 USD
76983-272 76983-274
2,5-Dichlorothiazole-4-carbonitrile 98%
2,5-Dichlorothiazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₄Cl₂N₂S
MW: 179.03 g/mol
MDL Number: MFCD16653247
CAS Number: 127426-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR