5-Chloro-1-methyl-1H-indole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A121228-1G A121228-250MG A121228-5G
76995-836EA 75 USD
76995-836 77880-984 77880-985
5-Chloro-1-methyl-1H-indole-2-carboxylic acid 98%
5-Chloro-1-methyl-1H-indole-2-carboxylic acid
Formula: C₁₀H₈ClNO₂
MDL Number: MFCD03848205
CAS Number: 59908-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR