1-Boc-4-(aminomethyl)-4-hydroxypiperidine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate, 1-Boc-4-(Aminomethyl)-4-hydroxypiperidine

A120997-1G A120997-250MG A120997-5G A120997-25G
77026-418EA 22 USD
77026-418 77026-420 77571-256 77871-534
1-Boc-4-(aminomethyl)-4-hydroxypiperidine 95%
1-Boc-4-(aminomethyl)-4-hydroxypiperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
MW: 230.3 g/mol
MDL Number: MFCD11579803
CAS Number: 392331-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR