Methyl 4-amino-2,5-dimethylbenzoate 95%

Supplier: Ambeed
Warning

A120984-10G A120984-250MG A120984-5G A120984-1G
77026-414EA 571.83 USD
77026-414 77026-416 77571-254 77666-736
Methyl 4-amino-2,5-dimethylbenzoate 95%
Methyl 4-amino-2,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
MDL Number: MFCD09839220
CAS Number: 21339-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR