2-Ethynyl-5-methoxypyridine 95%
Supplier: Ambeed
|
Warning
|
A120902-100MG
A120902-1G
A120902-5G
76983-262EA
30.77
USD
76983-262
77592-356
77683-086
2-Ethynyl-5-methoxypyridine 95%
2-Ethynyl-5-methoxypyridine
Precaución: For research use only.
Formula:
C₈H₇NO MW: 133.15 g/mol |
MDL Number:
MFCD13189716 CAS Number: 1196155-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...