Methyl-2,3-dichloroisonicotinate 95%

Supplier: Ambeed
Warning

A120876-1G A120876-250MG A120876-100MG
76995-826EA 55 USD
76995-826 76995-828 77880-979
Methyl-2,3-dichloroisonicotinate 95%
Methyl 2,3-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD09951946
CAS Number: 603124-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR