(R)-[(1-Phenylethyl)amino]acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((1-Phenylethyl)amino)acetic acid

A120845-10G A120845-1G A120845-250MG A120845-5G
76995-818EA 87 USD
76995-818 76995-820 76995-822 76995-824
(R)-[(1-Phenylethyl)amino]acetic acid 97%
(R)-[(1-Phenylethyl)amino]acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
MDL Number: MFCD11656779
CAS Number: 78397-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR