2-Amino-4-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A120829-250MG A120829-1G
77026-406EA 14 USD
77026-406 77871-532
2-Amino-4-fluorobenzaldehyde 98%
2-Amino-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD10000632
CAS Number: 152367-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR