cis-3,4-Difluoropyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (3S,4R)-rel-3,4-Difluoropyrrolidine hydrochloride

A120803-1G A120803-100MG
77009-426EA 233.4 USD
77009-426 77683-083
cis-3,4-Difluoropyrrolidine hydrochloride 97%
cis-3,4-Difluoropyrrolidine hydrochloride

Precaución: For research use only.

Formula: C₄H₈ClF₂N
MW: 143.56 g/mol
MDL Number: MFCD20230545
CAS Number: 869481-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR