4-Bromo-2-fluoro-6-methoxybenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-fluoro-6-methoxyphenylboronic acid, (4-Bromo-2-fluoro-6-methoxyphenyl)boronic acid, 2-Borono-5-bromo-3-fluoroanisole

A120792-1G A120792-250MG A120792-5G
77026-400EA 27.59 USD
77026-400 77026-402 77026-404
4-Bromo-2-fluoro-6-methoxybenzeneboronic acid 97%
4-Bromo-2-fluoro-6-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₃
MW: 248.842 g/mol
MDL Number: MFCD09475882
CAS Number: 957035-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR