3-Aminocyclohexanol 97%

Supplier: Ambeed
Danger

A120648-10G A120648-1G A120648-250MG A120648-25G A120648-5G A120648-100G
76983-230EA 49 USD
76983-230 76983-232 76983-234 76983-236 76983-238 77889-831
3-Aminocyclohexanol 97%
3-Aminocyclohexanol

Precaución: for Research Use Only

Formula: C6H13NO
MDL Number: MFCD03790923
CAS Number: 6850-39-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR