tert-Butyl (3S)-3-(methylamino)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 3-(methylamino)pyrrolidine-1-carboxylate

A120604-1G A120604-250MG A120604-25G A120604-10G A120604-5G
77026-394EA 39.26 USD
77026-394 77026-396 77026-398 77592-342 77599-736
tert-Butyl (3S)-3-(methylamino)pyrrolidine-1-carboxylate 97%
tert-Butyl (3S)-3-(methylamino)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.278 g/mol
MDL Number: MFCD09752962
CAS Number: 147081-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR