4-Bromo-5-fluoro-1H-indole 95%

Supplier: Ambeed
Warning

A120400-250MG A120400-50MG A120400-1G A120400-100MG
76983-216EA 172.93 USD
76983-216 76983-218 77571-218 77611-018
4-Bromo-5-fluoro-1H-indole 95%
4-Bromo-5-fluoro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.03 g/mol
MDL Number: MFCD16609917
CAS Number: 1227493-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR