Dapagliflozin 97%

Supplier: Ambeed
Danger

A120381-100MG A120381-10G A120381-10MG A120381-1G A120381-250MG A120381-25G A120381-50MG A120381-5G A120381-5MG
77026-366EA 21 USD
77026-366 77026-368 77026-370 77026-372 77026-374 77026-376 77026-378 77026-380 77026-382
Dapagliflozin 97%
Dapagliflozin

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClO₆
MDL Number: MFCD13182359
CAS Number: 461432-26-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR