5-Fluoro-2-methoxybenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 5-Fluoro-2-methoxybenzene-1-sulphonyl chloride

A120369-25G A120369-5G A120369-1G
76995-778EA 45 USD
76995-778 76995-780 77880-975
5-Fluoro-2-methoxybenzenesulfonyl chloride 98%
5-Fluoro-2-methoxybenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆ClFO₃S
MDL Number: MFCD03618459
CAS Number: 67475-56-3
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR