2-{[(2-Ethylhexyl)oxy]carbonyl}benzoic acid 97%

Supplier: Ambeed
Warning

A120361-100MG A120361-1G A120361-250MG A120361-25G A120361-5G A120361-10G
77026-356EA 12 USD
77026-356 77026-358 77026-360 77026-362 77026-364 77871-525
2-{[(2-Ethylhexyl)oxy]carbonyl}benzoic acid 97%
2-{[(2-Ethylhexyl)oxy]carbonyl}benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₄
MDL Number: MFCD00041500
CAS Number: 4376-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR