Adenosine 5'-diphosphate disodium salt (ADP disodium salt) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate
A120314-10G
A120314-1G
A120314-250MG
A120314-5G
A120314-25G
A120314-100G
76995-770EA
46.68
USD
76995-770
76995-772
76995-774
76995-776
77549-412
77566-222
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) 97%
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Precaución: For research use only.
Formula:
C₁₀H₁₃N₅Na₂O₁₀P₂ MW: 471.167 g/mol |
MDL Number:
MFCD00066635 CAS Number: 16178-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...