N-((3R,4R)-1-Benzyl-4-methylpiperidin-3-yl)-N-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A120278-100MG A120278-1G A120278-250MG A120278-25G A120278-5G
77026-342EA 23 USD
77026-342 77026-344 77026-346 77026-348 77026-350
N-((3R,4R)-1-Benzyl-4-methylpiperidin-3-yl)-N-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine 95%
N-((3R,4R)-1-Benzyl-4-methylpiperidin-3-yl)-N-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₂₀H₂₅N₅
MDL Number: MFCD16877475
CAS Number: 477600-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR