5-Methoxypyrimidin-4-amine 98%
Supplier: Ambeed
|
Warning
|
A120252-250MG
A120252-5G
A120252-1G
77026-338EA
38.2
USD
77026-338
77026-340
77666-722
5-Methoxypyrimidin-4-amine 98%
5-Methoxypyrimidin-4-amine
Precaución: for Research Use Only
Formula:
C₅H₇N₃O MW: 125.13 g/mol |
MDL Number:
MFCD01646126 CAS Number: 695-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...