Fmoc-3-(2-naphthyl)-D-alanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-2-Nal-OH

A120226-1G A120226-25G A120226-100G A120226-5G A120226-10G
76995-758EA 24.41 USD
76995-758 76995-760 77549-410 77571-212 77599-730
Fmoc-3-(2-naphthyl)-D-alanine 97%
Fmoc-3-(2-naphthyl)-D-alanine

Precaución: for Research Use Only

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
MDL Number: MFCD00077044
CAS Number: 138774-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR