6-Bromo-3,4-dihydro-2H-isoquinolin-1-one 98%
Supplier: Ambeed
|
Warning
|
A120127-100MG
A120127-10G
A120127-1G
A120127-250MG
A120127-5G
A120127-25G
77009-396EA
9
USD
77009-396
77009-398
77009-400
77009-402
77009-404
77571-204
6-Bromo-3,4-dihydro-2H-isoquinolin-1-one 98%
6-Bromo-3,4-dihydro-2H-isoquinolin-1-one
Precaución: for Research Use Only
Formula:
C₉H₈BrNO |
MDL Number:
MFCD04114858 CAS Number: 147497-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...