4-Iodobenzo[d][1,3]dioxole 95%

Supplier: Ambeed
Warning

A120074-1G A120074-5G
77026-330EA 184.6 USD
77026-330 77026-332
4-Iodobenzo[d][1,3]dioxole 95%
4-Iodobenzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₇H₅IO₂
MW: 248.01 g/mol
MDL Number: MFCD13152331
CAS Number: 849517-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR