(R)-Benzyl 3-hydroxypiperidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A119916-10G
A119916-1G
76983-196EA
286.45
USD
76983-196
77571-200
(R)-Benzyl 3-hydroxypiperidine-1-carboxylate 97%
(R)-Benzyl 3-hydroxypiperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₃ MW: 235.28 g/mol |
MDL Number:
MFCD11112280 CAS Number: 100858-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...