4-Chloro-2-methoxypyrimidine 97%

Supplier: Ambeed
Warning

A119864-100MG A119864-10G A119864-1G A119864-250MG A119864-5G
76983-186EA 15.92 USD
76983-186 76983-188 76983-190 76983-192 76983-194
4-Chloro-2-methoxypyrimidine 97%
4-Chloro-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.5590 g/mol
MDL Number: MFCD09025734
CAS Number: 51421-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR