(4-(Methoxymethyl)cyclohexyl)methanol 98%

Supplier: Ambeed
Warning

A119857-100MG A119857-1G A119857-5G
76983-180EA 17 USD
76983-180 76983-182 76983-184
(4-(Methoxymethyl)cyclohexyl)methanol 98%
(4-(Methoxymethyl)cyclohexyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₈O₂
MDL Number: MFCD22571682
CAS Number: 98955-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR