6-Fluoro-1H-indazole-3-carbonitrile 95%

Supplier: Ambeed
Warning

A119800-100MG A119800-1G A119800-250MG
77009-388EA 50.93 USD
77009-388 77009-390 77009-392
6-Fluoro-1H-indazole-3-carbonitrile 95%
6-Fluoro-1H-indazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄FN₃
MW: 161.136 g/mol
MDL Number: MFCD07371570
CAS Number: 885278-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR