Boc-2-chloro-L-phenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-3-(2-chlorophenyl)propanoic acid, 2-Chloro-L-phenylalanine, N-BOC protected, Boc-Phe(2-Cl)-OH
A119703-1G
A119703-250MG
A119703-25G
A119703-5G
76995-732EA
18
USD
76995-732
76995-734
76995-736
76995-738
Boc-2-chloro-L-phenylalanine 97%
Boc-2-chloro-L-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₄H₁₈ClNO₄ |
MDL Number:
MFCD00672514 CAS Number: 114873-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...