3-Bromo-N-ethylbenzamide 98%

Supplier: Ambeed
Warning

A119701-5G A119701-1G A119701-250MG
76983-174EA 81.99 USD
76983-174 77889-825 77889-826
3-Bromo-N-ethylbenzamide 98%
3-Bromo-N-ethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MDL Number: MFCD01215180
CAS Number: 26819-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR