5-Bromo-2-(methylsulfonyl)pyrimidine 98%

Supplier: Ambeed
Warning

A119596-1G A119596-250MG A119596-5G
77009-378EA 24 USD
77009-378 77009-380 77009-382
5-Bromo-2-(methylsulfonyl)pyrimidine 98%
5-Bromo-2-(methylsulfonyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂S
MDL Number: MFCD00508247
CAS Number: 38275-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR