1,6-Dimethyl-1H-benzo[d]imidazole 95%

Supplier: Ambeed
Warning

A119595-1G A119595-250MG
76983-158EA 235 USD
76983-158 76983-160
1,6-Dimethyl-1H-benzo[d]imidazole 95%
1,6-Dimethyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MDL Number: MFCD08685462
CAS Number: 10394-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR