2-Chloro-5-methylnicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-methylpyridine-3-carboxylic acid

A119577-1G A119577-250MG A119577-5G A119577-25G A119577-100G
76983-150EA 11 USD
76983-150 76983-152 76983-154 77611-006 77889-824
2-Chloro-5-methylnicotinic acid 97%
2-Chloro-5-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MDL Number: MFCD07375371
CAS Number: 66909-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR