7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A119553-1G
A119553-250MG
A119553-5G
77009-372EA
145.35
USD
77009-372
77009-374
77009-376
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₁ClF₃N MW: 237.6500 g/mol |
MDL Number:
MFCD05861544 CAS Number: 220247-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...